Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM217A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010482.1 | XP_016865971.1 | 1, 2 | 333 | ||
GenBank mRNA | AK097426.1 | - | 1, 2 | 333 |