Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003491.3 | 5 | 6184 | |||
NR_033319.2 | 4 | 6058 | ||||
NR_033320.2 | 5 | 6110 | ||||
NR_033321.2 | 4 | 5984 | ||||
GenBank mRNA | AB263414.1 | - | 5 | 6145 | ||
AB263415.1 | - | 4 | 6019 | |||
AB263416.1 | - | 5 | 6071 | |||
AB263417.1 | - | 4 | 5945 | |||
AK127256.1 | - | 1 | 1297 |
Gene Symbol: LOC100129936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124820.1 | - | 1 | 173 |