Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: POLR2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713666.2 | XP_006713729.1 | 1, 2 | 433 | ||
XM_006713668.3 | XP_006713731.1 | 1, 2 | 442 | |||
XM_006713670.2 | XP_006713733.1 | 1, 2 | 433 | |||
XM_017006636.1 | XP_016862125.1 | 1, 2 | 433 | |||
XM_017006637.1 | XP_016862126.1 | 1, 2 | 433 | |||
GenBank mRNA | HY178178.1 | - | 1, 2 | 438 |