Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142964.1 | NP_001136436.1 | 2 | 1812 | ||
XM_011530373.2 | XP_011528675.1 | 3 | 1720 | |||
XM_017028939.1 | XP_016884428.1 | 4 | 1851 | |||
GenBank mRNA | AK095008.1 | - | 1 | 565 | ||
AK131457.1 | - | 4 | 1644 | |||
BC035403.1 | - | 2 | 1812 |