Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129890.1 | NP_001123362.1 | 6, 7 | 797 | ||
GenBank mRNA | AK130865.1 | - | 2, 3 | 303 |