Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129890.1 | NP_001123362.1 | 7, 8 | 975 | ||
GenBank mRNA | AK130865.1 | - | 3, 4 | 481 |