Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016848.5 | NP_058544.3 | 12 | 7375 | ||
XM_011518785.2 | XP_011517087.1 | 12 | 10917 | |||
XM_011518786.2 | XP_011517088.1 | 13 | 6766 | |||
GenBank mRNA | AK127258.1 | - | 1 | 1131 | ||
BC042431.1 | - | 1 | 1478 | |||
BX538228.1 | - | 1 | 2237 |
Gene Symbol: LOC100131540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129921.1 | - | 1 | 3533 |