Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027263.1 | 1 | 230 | |||
GenBank mRNA | AK314153.1 | - | 1 | 172 | ||
BC007251.1 | - | 1 | 93 | |||
BM745256.1 | - | 1 | 230 |
Gene Symbol: ARHGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267636.3 | XP_005267693.1 | 1 | 171 |