Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM196B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129891.1 | NP_001123363.1 | 3 | 2788 | ||
XM_011534415.2 | XP_011532717.1 | 3 | 2477 | |||
XM_017008908.1 | XP_016864397.1 | 3 | 2556 | |||
GenBank mRNA | AK055998.1 | - | 3 | 2788 |