Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121630.1 | 1 | 268 | |||
GenBank mRNA | DB177224.1 | - | 1 | 127 | ||
DB196812.1 | - | 1 | 268 |