Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195263.1 | NP_001182192.1 | 15 | 2412 | ||
XM_011540177.2 | XP_011538479.1 | 16 | 2572 | |||
XM_011540178.2 | XP_011538480.1 | 15 | 2384 | |||
GenBank mRNA | AK024422.1 | - | 11 | 2709 | ||
FJ617449.1 | - | 16 | 2290 |