Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM231B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310155.1 | NP_001297084.1 | 1 | 584 | ||
XM_016999988.1 | XP_016855477.1 | 2 | 3293 |