Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000800.4 | NP_000791.1 | 1 | 415 | ||
NM_033136.3 | NP_149127.1 | 1 | 415 | |||
GenBank mRNA | DA126618.1 | - | 1 | 407 | ||
DA189432.1 | - | 1 | 45 | |||
DA326831.1 | - | 1 | 73 | |||
DB092576.1 | - | 1 | 396 | |||
M74028.1 | - | 1 | 347 |