Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNUPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042581.1 | NP_001036046.1 | 9 | 1359 | ||
NM_001042588.1 | NP_001036053.1 | 9 | 1164 | |||
NM_005701.3 | NP_005692.1 | 9 | 1464 | |||
GenBank mRNA | AF039029.1 | - | 9 | 1109 | ||
AK054641.1 | - | 10 | 1597 | |||
AK289475.1 | - | 9 | 1160 | |||
BC004203.2 | - | 9 | 1095 | 3605328 | ||
CR456811.1 | - | 8 | 1019 |