Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032381.1 | NP_001027553.1 | 4, 5 | 407 | ||
NM_001032382.1 | NP_001027554.1 | 4, 5 | 475 | |||
NM_001032383.1 | NP_001027555.1 | 4, 5 | 486 | |||
NM_001032384.1 | NP_001027556.1 | 4, 5 | 395 | |||
NM_001167989.1 | NP_001161461.1 | 4, 5 | 407 | |||
NM_001167990.1 | NP_001161462.1 | 4, 5 | 365 | |||
NM_005710.2 | NP_005701.1 | 3, 4 | 540 | |||
XM_005272571.3 | XP_005272628.1 | 4, 5 | 470 | |||
XM_011543884.2 | XP_011542186.1 | 4, 5 | 545 | |||
XM_017029207.1 | XP_016884696.1 | 4, 5 | 536 | |||
GenBank mRNA | AB016533.1 | - | 4, 5 | 456 | ||
AB041834.1 | - | 3, 4 | 286 | |||
AB041835.1 | - | 3, 4 | 286 | |||
AJ005893.1 | - | 3, 4 | 543 | |||
AJ242829.1 | - | 4, 5 | 346 | |||
AJ973594.1 | - | 3, 4 | 301 | |||
AJ973595.1 | - | 3, 4 | 301 | |||
AJ973596.1 | - | 301 | ||||
AJ973597.1 | - | 3, 4 | 301 | |||
AJ973598.1 | - | 3, 4 | 301 | |||
AJ973599.1 | - | 3, 4 | 301 | |||
AJ973600.1 | - | 3, 4 | 301 | |||
AJ973601.1 | - | 4, 5 | 388 | |||
AJ973602.1 | - | 3, 4 | 605 | |||
AJ973603.1 | - | 4, 5 | 388 | |||
AJ973604.1 | - | 3, 4 | 305 | |||
AJ973606.1 | - | 3, 4 | 301 | |||
BC012358.1 | - | 4, 5 | 367 | 4662649 | ||
BE385548.1 | - | 2, 3 | 221 | |||
BE396796.1 | - | 4, 5 | 387 | |||
BI818141.1 | - | 4, 5 | 369 | |||
BX362311.2 | - | 4, 5 | 444 | |||
BX369697.2 | - | 4, 5 | 627 | |||
CD051018.1 | - | 4, 5 | 386 | |||
CD365745.1 | - | 3, 4 | 588 | |||
CN292761.1 | - | 4, 5 | 475 | |||
CN292769.1 | - | 4, 5 | 393 | |||
KU178500.1 | - | 3, 4 | 286 | |||
KU178501.1 | - | 4, 5 | 334 |