Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284501.1 | NP_001271430.1 | 6 | 845 | ||
NM_001284502.1 | NP_001271431.1 | 4 | 456 | |||
NM_012225.3 | NP_036357.1 | 5 | 655 | |||
NR_104318.1 | 4 | 456 | ||||
XM_005255027.2 | XP_005255084.1 | 5 | 1122 | |||
XM_005255030.1 | XP_005255087.1 | 4 | 393 | |||
XM_011522338.2 | XP_011520640.1 | 4 | 590 | |||
XM_017022831.1 | XP_016878320.1 | 5 | 789 | |||
XM_017022832.1 | XP_016878321.1 | 4 | 923 | |||
GenBank mRNA | AF118394.1 | - | 5 | 620 | ||
AK001023.1 | - | 5 | 631 | |||
AK093593.1 | - | 3 | 1468 | |||
AK300696.1 | - | 4 | 389 | |||
AK315611.1 | - | 5 | 596 | |||
BC002768.2 | - | 5 | 612 | 3633393 | ||
BC008005.1 | - | 5 | 637 | 3507768 | ||
BI196883.1 | - | 4 | 381 | |||
BX648907.1 | - | 3 | 1423 |