Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324069.1 | NP_001310998.1 | 12 | 1206 | ||
NM_001324070.1 | NP_001310999.1 | 8 | 948 | |||
NM_001324071.1 | NP_001311000.1 | 9 | 982 | |||
NM_001324072.1 | NP_001311001.1 | 11 | 983 | |||
NM_001324074.1 | NP_001311003.1 | 10 | 1085 | |||
NM_001324075.1 | NP_001311004.1 | 11 | 1117 | |||
NM_001324077.1 | NP_001311006.1 | 11 | 985 | |||
NM_001324078.1 | NP_001311007.1 | 10 | 1083 | |||
NM_001799.3 | NP_001790.1 | 11 | 1119 | |||
NR_136690.1 | 10 | 1000 | ||||
XM_011543094.2 | XP_011541396.1 | 8 | 789 | |||
GenBank mRNA | AK026509.1 | - | 11 | 1004 | ||
BC000834.2 | - | 11 | 958 | 3455907 | ||
BC005298.1 | - | 11 | 1026 | 3997991 | ||
CA305960.1 | - | 2 | 300 | |||
CA437275.1 | - | 2 | 412 | |||
KU177970.1 | - | 11 | 938 | |||
KU177971.1 | - | 10 | 902 | |||
L20320.1 | - | 12 | 1113 | |||
X77303.1 | - | 11 | 981 | |||
X77743.1 | - | 11 | 972 | |||
X79193.1 | - | 11 | 1028 | |||
Y13120.1 | - | 11 | 1041 |