Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278911.1 | NP_001265840.1 | 3 | 519 | ||
NM_005832.4 | NP_005823.1 | 4 | 596 | |||
NM_181361.2 | NP_852006.1 | 3 | 648 | |||
XM_011512325.2 | XP_011510627.1 | 4 | 629 | |||
GenBank mRNA | AA904191.1 | - | 4 | 434 | ||
AB193524.1 | - | 3 | 110 | |||
AB193528.1 | - | 2, 3 | 167 | |||
AB193530.1 | - | 2 | 167 | |||
AF099137.1 | - | 3 | 312 | |||
AF209747.1 | - | 3 | 519 | |||
AK298291.1 | - | 3 | 488 | |||
BC017825.1 | - | 3 | 510 | 4657825 | ||
BG701449.1 | - | 3 | 657 |