Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005876.4 | NP_005867.3 | 27 | 5417 | ||
XM_005246237.2 | XP_005246294.1 | 29 | 5145 | |||
XM_005246239.2 | XP_005246296.1 | 29 | 4960 | |||
XM_005246240.2 | XP_005246297.1 | 22 | 3162 | |||
XM_005246241.1 | XP_005246298.1 | 22 | 3056 | |||
XM_005246242.4 | XP_005246299.1 | 22 | 3181 | |||
XM_006712189.3 | XP_006712252.1 | 28 | 5150 | |||
XM_006712193.3 | XP_006712256.1 | 22 | 6281 | |||
XM_011510479.2 | XP_011508781.1 | 28 | 5583 | |||
XM_011510483.2 | XP_011508785.2 | 28 | 5180 | |||
XM_017003157.1 | XP_016858646.1 | 28 | 5180 | |||
XM_017003158.1 | XP_016858647.1 | 24 | 4756 | |||
XM_017003159.1 | XP_016858648.1 | 25 | 4939 | |||
XM_017003160.1 | XP_016858649.1 | 12 | 1352 | |||
GenBank mRNA | AB037718.1 | - | 18 | 2474 | ||
AY603755.1 | - | 26 | 5030 |