Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNIP1 SIRNA Assays ›
Gene Symbol:
TNIP1
Gene Name:
TNFAIP3 interacting protein 1
Gene Aliases:
ABIN-1, NAF1, VAN, nip40-1
Chromosome Location:
Chr.5: 151029943 - 151087660 on Build GRCh38
UniGene ID:
Hs.355141
Species:
Human
siRNA ID
HSS173556
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001252385.1
NP_001239314.1
3
430
NM_001252386.1
NP_001239315.1
2
258
NM_001252390.1
NP_001239319.1
3
295
NM_001252391.1
NP_001239320.1
3
327
NM_001252392.1
NP_001239321.1
3
327
NM_001252393.1
NP_001239322.1
3
430
NM_001258454.1
NP_001245383.1
3
430
NM_001258455.1
NP_001245384.1
2
225
NM_001258456.1
NP_001245385.1
2
225
NM_006058.4
NP_006049.3
3
430
XM_005268355.1
XP_005268412.1
3
343
XM_006714751.1
XP_006714814.1
3
343
XM_006714752.2
XP_006714815.1
3
343
XM_017008945.1
XP_016864434.1
3
343
XM_017008946.1
XP_016864435.1
3
309
XM_017008947.1
XP_016864436.1
4
812
XM_017008948.1
XP_016864437.1
2
604
XM_017008949.1
XP_016864438.1
3
343
XM_017008950.1
XP_016864439.1
3
343
XM_017008951.1
XP_016864440.1
2
604
GenBank mRNA
AB177543.1
-
2
189
AB177544.1
-
2
189
AB252970.1
-
3
299
AB252971.1
-
3
299
AB252972.1
-
3
420
AB252973.1
-
3
420
AB252974.1
-
2
127
AB252975.1
-
2
248
AB252976.1
-
1
53
AB252977.1
-
1
53
AB252978.1
-
1
53
AB252979.1
-
1
53
AB252980.1
-
1
53
AB252981.1
-
1
53
AB252982.1
-
3
299
AJ011895.1
-
3
299
AJ011896.1
-
3
299
AK074400.1
-
3
361
AK290999.1
-
3
296
AK298685.1
-
2
252
AK299975.1
-
2
190
AK303573.1
-
3
380
AY012155.1
-
3
446
BC012133.1
-
3
336
4549542
BC014008.1
-
3
277
4040118
BP254790.1
-
3
420
BX640647.1
-
2
179
D30755.2
-
2
189
DA643531.1
-
2
155
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MyD88-dependent toll-like receptor signaling pathway
translation
proteolysis
defense response
inflammatory response
leukocyte cell-cell adhesion
glycoprotein biosynthetic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of viral genome replication
positive regulation of transcription from RNA polymerase II promoter
positive regulation of inflammatory response
negative regulation of ERK1 and ERK2 cascade
modulation by symbiont of host I-kappaB kinase/NF-kappaB cascade
thiol-dependent ubiquitin-specific protease activity
protein binding
mitogen-activated protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-gnqd7:80/10.239.161.169:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline