Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006084.4 | NP_006075.3 | 3 | 397 | ||
GenBank mRNA | AK295467.1 | - | 3 | 290 | ||
AK308347.1 | - | 3 | 320 | |||
BC035716.2 | - | 2 | 273 | 5588434 | ||
BI919023.1 | - | 3 | 584 | |||
M87503.1 | - | 3 | 304 | |||
U92994.1 | - | 1 | 60 |