Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006336.3 | NP_006327.2 | 2 | 589 | ||
XM_005251645.2 | XP_005251702.1 | 2 | 381 | |||
XM_005251646.2 | XP_005251703.1 | 2 | 584 | |||
XM_011518122.2 | XP_011516424.1 | 2 | 391 | |||
XM_017014185.1 | XP_016869674.1 | 2 | 997 | |||
XM_017014186.1 | XP_016869675.1 | 2 | 378 | |||
XM_017014187.1 | XP_016869676.1 | 2 | 997 | |||
XM_017014188.1 | XP_016869677.1 | 2 | 365 | |||
XM_017014189.1 | XP_016869678.1 | 2 | 584 | |||
XM_017014190.1 | XP_016869679.1 | 2 | 378 | |||
GenBank mRNA | AK314911.1 | - | 2 | 337 | ||
BC033884.1 | - | 2 | 558 | |||
BC052563.1 | - | 2 | 574 | 6303965 | ||
X99802.1 | - | 1 | 179 |