Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006111.2 | NP_006102.2 | 2, 3 | 588 | ||
GenBank mRNA | AK054673.1 | - | 2, 3 | 588 | ||
AK225457.1 | - | 2, 3 | 239 | |||
AK314462.1 | - | 2, 3 | 590 | |||
BC001918.1 | - | 2, 3 | 210 | 2820744 | ||
D16294.1 | - | 2, 3 | 239 |