Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006361.5 | NP_006352.2 | 2 | 977 | ||
GenBank mRNA | AK225234.1 | - | 2 | 921 | ||
AK313266.1 | - | 2 | 920 | |||
AY937237.1 | - | 2 | 986 | |||
BC007092.1 | - | 2 | 907 | 4250486 | ||
BC070233.1 | - | 2 | 912 | 6668163 | ||
BT007410.1 | - | 2 | 821 | |||
U57052.1 | - | 2 | 875 | |||
U81599.1 | - | 2 | 997 |