Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105530.1 | NP_001099000.1 | 4 | 470 | ||
NM_006367.3 | NP_006358.1 | 4 | 488 | |||
XM_005270368.1 | XP_005270425.1 | 4 | 328 | |||
XM_011540509.1 | XP_011538811.1 | 5 | 881 | |||
XM_011540510.1 | XP_011538812.1 | 5 | 878 | |||
XM_011540511.1 | XP_011538813.1 | 4 | 312 | |||
XM_011540512.1 | XP_011538814.1 | 4 | 431 | |||
XM_011540513.2 | XP_011538815.1 | 4 | 323 | |||
XM_011540514.1 | XP_011538816.1 | 5 | 438 | |||
XM_011540515.1 | XP_011538817.1 | 5 | 878 | |||
XM_017000067.1 | XP_016855556.1 | 4 | 396 | |||
XM_017000068.1 | XP_016855557.1 | 5 | 440 | |||
XM_017000069.1 | XP_016855558.1 | 5 | 651 | |||
XM_017000070.1 | XP_016855559.1 | 4 | 670 | |||
XM_017000071.1 | XP_016855560.1 | 4 | 628 | |||
XM_017000072.1 | XP_016855561.1 | 4 | 564 | |||
XM_017000073.1 | XP_016855562.1 | 4 | 385 | |||
GenBank mRNA | AK096193.1 | - | 4 | 322 | ||
AK222513.1 | - | 4 | 329 | |||
AK295396.1 | - | 5 | 250 | |||
AK298094.1 | - | 4 | 168 | |||
AK298110.1 | - | 4 | 272 | |||
AK315731.1 | - | 4 | 375 | |||
BC013963.2 | - | 4 | 305 | 2963922 | ||
BC095440.1 | - | 4 | 310 | 30370256 | ||
BT007152.1 | - | 3 | 259 | |||
CR457409.1 | - | 3 | 259 | |||
L12168.1 | - | 3 | 298 | |||
M98474.1 | - | 4 | 329 |