Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAP1 SIRNA Assays ›
Gene Symbol:
CAP1
Gene Name:
adenylate cyclase associated protein 1
Gene Aliases:
CAP, CAP1-PEN
Chromosome Location:
Chr.1: 40040070 - 40072649 on Build GRCh38
UniGene ID:
Hs.370581
Species:
Human
siRNA ID
HSS173677
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105530.1
NP_001099000.1
5
609
NM_006367.3
NP_006358.1
5
627
XM_005270368.1
XP_005270425.1
5
467
XM_011540509.1
XP_011538811.1
6
1020
XM_011540510.1
XP_011538812.1
6
1017
XM_011540511.1
XP_011538813.1
5
451
XM_011540512.1
XP_011538814.1
5
570
XM_011540513.2
XP_011538815.1
5
462
XM_011540514.1
XP_011538816.1
6
577
XM_011540515.1
XP_011538817.1
6
1017
XM_017000067.1
XP_016855556.1
5
535
XM_017000068.1
XP_016855557.1
6
579
XM_017000069.1
XP_016855558.1
6
790
XM_017000070.1
XP_016855559.1
5
809
XM_017000071.1
XP_016855560.1
5
767
XM_017000072.1
XP_016855561.1
5
703
XM_017000073.1
XP_016855562.1
5
524
GenBank mRNA
AK096193.1
-
5
461
AK222513.1
-
5
468
AK295396.1
-
6
389
AK297829.1
-
5
336
AK298094.1
-
5
307
AK298110.1
-
5
411
AK300861.1
-
4
306
AK315731.1
-
5
514
BC013963.2
-
5
444
2963922
BC095440.1
-
5
449
30370256
BT007152.1
-
4
398
CR457409.1
-
4
398
L12168.1
-
4
437
M98474.1
-
5
468
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
ameboidal-type cell migration
receptor-mediated endocytosis
establishment or maintenance of cell polarity
signal transduction
activation of adenylate cyclase activity
actin polymerization or depolymerization
actin binding
adenylate cyclase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0