Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006600.3 | NP_006591.1 | 6 | 850 | ||
XM_011540529.1 | XP_011538831.1 | 6 | 750 | |||
XM_011540530.1 | XP_011538832.1 | 6 | 988 | |||
XM_017000094.1 | XP_016855583.1 | 7 | 921 | |||
XM_017000095.1 | XP_016855584.1 | 7 | 1020 | |||
XM_017000096.1 | XP_016855585.1 | 7 | 1030 | |||
GenBank mRNA | AB019408.1 | - | 6 | 797 | ||
AF086922.1 | - | 6 | 813 | |||
AF100760.1 | - | 6 | 813 | |||
AF125465.1 | - | 6 | 821 | |||
AF130736.1 | - | 6 | 764 | |||
AF241788.1 | - | 6 | 798 | |||
AL136725.1 | - | 6 | 826 | |||
BC002399.2 | - | 6 | 762 | 2822167 | ||
BC003132.1 | - | 6 | 770 | 2958251 | ||
BC006147.1 | - | 6 | 796 | 3954220 | ||
BC007280.1 | - | 6 | 839 | 3343588 | ||
BC015153.1 | - | 6 | 838 | 3883328 | ||
BC021139.1 | - | 6 | 811 | 4871007 |