Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCFL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301726.1 | NP_001288655.1 | 4 | 1309 | ||
NM_006602.3 | NP_006593.2 | 4 | 1312 | |||
XM_005260185.3 | XP_005260242.1 | 4 | 1216 | |||
XM_011528497.2 | XP_011526799.1 | 4 | 1216 | |||
XM_011528498.2 | XP_011526800.1 | 4 | 1216 | |||
XM_011528499.1 | XP_011526801.1 | 4 | 1216 | |||
GenBank mRNA | AB012124.1 | - | 4 | 1173 | ||
AF070992.1 | - | 4 | 1150 | |||
AJ271337.1 | - | 3 | 1021 | |||
BC046933.1 | - | 4 | 1313 | 5549053 |