Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013436.4 | NP_038464.1 | 15 | 2218 | ||
NM_205842.2 | NP_995314.1 | 16 | 2236 | |||
XM_006712200.3 | XP_006712263.1 | 16 | 1722 | |||
XM_006712201.3 | XP_006712264.1 | 15 | 1706 | |||
GenBank mRNA | AB011159.1 | - | 16 | 2236 | ||
AB014509.1 | - | 15 | 1478 | |||
AK292914.1 | - | 15 | 1708 | |||
BC015025.1 | - | 15 | 1730 | 3907646 | ||
BT007033.1 | - | 15 | 1459 |