Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP19 SIRNA Assays ›
Gene Symbol:
USP19
Gene Name:
ubiquitin specific peptidase 19
Gene Aliases:
ZMYND9
Chromosome Location:
Chr.3: 49108046 - 49120938 on Build GRCh38
UniGene ID:
Hs.255596
Species:
Human
siRNA ID
HSS173936
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199160.1
NP_001186089.1
24
3931
NM_001199161.1
NP_001186090.1
24
3937
NM_001199162.1
NP_001186091.1
24
3901
NM_006677.2
NP_006668.1
23
3628
XM_005264823.2
XP_005264880.1
24
3938
XM_005264824.2
XP_005264881.1
24
3957
XM_005264825.2
XP_005264882.1
24
3932
XM_005264826.2
XP_005264883.1
24
3932
XM_005264827.2
XP_005264884.1
24
3893
XM_005264829.2
XP_005264886.1
24
3938
XM_005264830.2
XP_005264887.1
23
3635
XM_005264831.2
XP_005264888.1
23
3584
XM_006712946.2
XP_006713009.1
24
3935
XM_006712947.2
XP_006713010.1
24
3884
XM_006712948.2
XP_006713011.1
24
3926
XM_006712949.2
XP_006713012.1
24
3788
XM_006712950.2
XP_006713013.1
24
3887
XM_006712951.2
XP_006713014.1
24
3884
XM_006712952.2
XP_006713015.1
24
3881
XM_006712953.1
XP_006713016.1
24
3852
XM_017005615.1
XP_016861104.1
24
3929
XM_017005616.1
XP_016861105.1
24
3923
XM_017005617.1
XP_016861106.1
24
3887
XM_017005618.1
XP_016861107.1
24
3932
XM_017005619.1
XP_016861108.1
24
3929
XM_017005620.1
XP_016861109.1
24
3923
XM_017005621.1
XP_016861110.1
24
3782
XM_017005622.1
XP_016861111.1
24
3893
XM_017005623.1
XP_016861112.1
24
3776
XM_017005624.1
XP_016861113.1
24
3776
XM_017005625.1
XP_016861114.1
23
3629
XM_017005626.1
XP_016861115.1
23
3642
XM_017005627.1
XP_016861116.1
23
3590
XM_017005628.1
XP_016861117.1
23
3584
XM_017005629.1
XP_016861118.1
23
3699
XM_017005630.1
XP_016861119.1
23
3635
XM_017005631.1
XP_016861120.1
23
3629
XM_017005632.1
XP_016861121.1
23
3629
XM_017005633.1
XP_016861122.1
23
3623
XM_017005634.1
XP_016861123.1
23
3590
XM_017005635.1
XP_016861124.1
17
2366
GenBank mRNA
AB020698.1
-
23
3470
AK294756.1
-
24
3824
AK300425.1
-
24
3755
BC014934.1
-
19
2909
BC048269.1
-
5
776
BC065909.1
-
12
1708
BC082241.1
-
21
3004
BC142660.1
-
24
3807
BC142727.1
-
24
3813
BC146752.1
-
23
3470
9007186
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
ER-associated ubiquitin-dependent protein catabolic process
regulation of protein stability
response to endoplasmic reticulum stress
negative regulation of skeletal muscle tissue development
protein stabilization
protein K48-linked deubiquitination
positive regulation of cell cycle process
regulation of cellular response to hypoxia
negative regulation of proteasomal protein catabolic process
regulation of ERAD pathway
thiol-dependent ubiquitin-specific protease activity
protein binding
ubiquitin protein ligase binding
metal ion binding
Hsp90 protein binding
Lys48-specific deubiquitinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0