Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016540.3 | NP_057624.3 | 3 | 790 | ||
XM_011542559.2 | XP_011540861.1 | 2 | 724 | |||
GenBank mRNA | AB040973.1 | - | 3 | 623 | ||
AF236081.1 | - | 3 | 618 | |||
AK315161.1 | - | 3 | 790 | |||
BC067473.1 | - | 3 | 618 | 6971838 | ||
BC067474.1 | - | 3 | 618 | 6971839 | ||
EU432120.1 | - | 3 | 618 |
Set Membership: |
![]() |