Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014599.5 | NP_055414.2 | 4 | 702 | ||
NM_177433.2 | NP_803182.1 | 4 | 810 | |||
NM_201222.2 | NP_957516.1 | 4 | 770 | |||
GenBank mRNA | AF126181.1 | - | 4 | 646 | ||
AF128527.1 | - | 4 | 646 | |||
AF128528.1 | - | 4 | 646 | |||
AJ293618.1 | - | 3 | 589 | |||
AK092463.1 | - | 5 | 608 | |||
AK098645.1 | - | 5 | 555 | |||
BC000304.2 | - | 4 | 664 | 2820442 | ||
BC091503.1 | - | 4 | 642 | 30529545 | ||
BM803170.1 | - | 676 | ||||
BQ423605.1 | - | 4 | 671 | |||
BX647995.1 | - | 4 | 747 | |||
U92544.1 | - | 4 | 643 |