Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014267.5 | NP_055082.1 | 3 | 337 | ||
XM_017017143.1 | XP_016872632.1 | 3 | 2596 | |||
GenBank mRNA | AK057169.1 | - | 1 | 1405 | ||
AK315382.1 | - | 3 | 291 | |||
BC007103.1 | - | 3 | 291 | 4281282 | ||
BC016352.1 | - | 3 | 295 | 4082845 | ||
BC020937.1 | - | 3 | 291 | 4281930 | ||
BC103747.1 | - | 3 | 318 | 6151644 | ||
BP210918.1 | - | 4 | 375 | |||
BP226882.1 | - | 3 | 537 | |||
BU661194.1 | - | 4 | 386 | |||
CR456882.1 | - | 3 | 159 | |||
U51678.1 | - | 3 | 194 |