Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM107A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076778.2 | NP_001070246.1 | 2 | 613 | ||
NM_001282713.1 | NP_001269642.1 | 2 | 413 | |||
NM_001282714.1 | NP_001269643.1 | 3 | 861 | |||
NM_007177.3 | NP_009108.1 | 3 | 711 | |||
GenBank mRNA | AB023811.1 | - | 2 | 193 | ||
AF035283.1 | - | 2 | 613 | |||
AF089853.1 | - | 2 | 704 | |||
AF089854.1 | - | 3 | 802 | |||
AK054720.1 | - | 3 | 861 | |||
AK055443.1 | - | 2 | 237 | |||
AK128791.1 | - | 2 | 240 | |||
AK316450.1 | - | 2 | 268 | |||
AL050264.1 | - | 2 | 226 | |||
BC010561.1 | - | 3 | 312 | 4152118 | ||
BP212425.1 | - | 3 | 418 | |||
BX645008.1 | - | 2 | 423 | |||
CR457133.1 | - | 1 | 152 | |||
EU569833.1 | - | 2 | 268 |