Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172648.3 | NP_001166119.1 | 4 | 883 | ||
NM_001172649.3 | NP_001166120.1 | 4 | 1130 | |||
NM_001291106.1 | NP_001278035.1 | 4 | 1130 | |||
NM_001291107.1 | NP_001278036.1 | 4 | 1127 | |||
NM_007185.6 | NP_009116.3 | 4 | 1130 | |||
XM_006711139.2 | XP_006711202.1 | 5 | 358 | |||
XM_006711141.2 | XP_006711204.1 | 5 | 355 | |||
XM_011509111.2 | XP_011507413.1 | 4 | 978 | |||
XM_011509112.2 | XP_011507414.1 | 3 | 221 | |||
XM_011509114.2 | XP_011507416.1 | 3 | 189 | |||
XM_017000145.1 | XP_016855634.1 | 3 | 196 | |||
XM_017000146.1 | XP_016855635.1 | 3 | 193 | |||
GenBank mRNA | AB209264.1 | - | 4 | 883 | ||
AF329264.1 | - | 4 | 330 | |||
AL519587.3 | - | 5 | 362 | |||
AY165003.1 | - | 4 | 1120 | |||
BC004145.2 | - | 5 | 305 | |||
BC041088.1 | - | 4 | 362 | |||
BC052491.1 | - | 4 | 1104 | 4820737 | ||
BC068008.1 | - | 3 | 157 | |||
BC104758.1 | - | 4 | 393 | 8143761 | ||
BC143226.1 | - | 4 | 390 | 9051727 |