Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271854.1 | NP_001258783.1 | 4 | 1767 | ||
NM_001271855.1 | NP_001258784.1 | 3 | 928 | |||
NM_007223.2 | NP_009154.1 | 3 | 1770 | |||
XM_011521167.2 | XP_011519469.1 | 5 | 1418 | |||
XM_011521168.2 | XP_011519470.1 | 4 | 1909 | |||
XM_017021873.1 | XP_016877362.1 | 4 | 1334 | |||
XM_017021874.1 | XP_016877363.1 | 4 | 1990 | |||
XM_017021875.1 | XP_016877364.1 | 5 | 2093 | |||
XM_017021876.1 | XP_016877365.1 | 5 | 1502 | |||
XM_017021877.1 | XP_016877366.1 | 7 | 1702 | |||
XM_017021878.1 | XP_016877367.1 | 2 | 1558 | |||
GenBank mRNA | AK302715.1 | - | 3 | 928 | ||
BC067106.1 | - | 4 | 1100 | 6645890 | ||
D38449.1 | - | 3 | 1770 | |||
JQ308174.1 | - | 3 | 904 |
Set Membership: |
G-protein coupled receptor |