Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284341.1 | NP_001271270.1 | 6 | 1172 | ||
NM_001284343.1 | NP_001271272.1 | 2 | 250 | |||
NM_007270.4 | NP_009201.2 | 5 | 1013 | |||
XM_011515115.2 | XP_011513417.1 | 5 | 1013 | |||
GenBank mRNA | AF089745.1 | - | 5 | 684 | ||
AK075331.1 | - | 5 | 977 | |||
AK128597.1 | - | 6 | 954 | |||
AK293440.1 | - | 2 | 228 | |||
AK293954.1 | - | 4 | 586 | |||
AK294266.1 | - | 4 | 797 | |||
AK300328.1 | - | 6 | 1135 | |||
BC101723.1 | - | 5 | 941 | 8069229 | ||
BC112053.1 | - | 5 | 941 | 8327521 | ||
BQ688269.1 | - | 2 | 250 |