Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFT2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145169.2 | NP_660152.1 | 2 | 227 | ||
NR_130112.1 | 3 | 250 | ||||
NR_130113.1 | 3 | 250 | ||||
NR_130114.1 | 3 | 241 | ||||
GenBank mRNA | AF041429.1 | - | 2 | 161 | ||
AW964442.1 | - | 3 | 172 | |||
BC018969.1 | - | 2 | 132 | 3959886 | ||
BC028388.1 | - | 3 | 194 | |||
BF223532.1 | - | 3 | 145 | |||
BG706927.1 | - | 2 | 216 | |||
BI597156.1 | - | 3 | 212 | |||
DB476616.1 | - | 3 | 250 |