Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBPL9 SIRNA Assays ›
Gene Symbol:
OSBPL9
Gene Name:
oxysterol binding protein like 9
Gene Aliases:
ORP-9, ORP9
Chromosome Location:
Chr.1: 51616874 - 51789219 on Build GRCh38
UniGene ID:
Hs.21938
Species:
Human
siRNA ID
HSS174371
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBPL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024586.5
NP_078862.4
12
1069
NM_148904.3
NP_683702.1
8
786
NM_148905.3
NP_683703.1
7
690
NM_148906.2
NP_683704.2
11
1018
NM_148907.2
NP_683705.1
8
663
NM_148908.3
NP_683706.3
11
1030
NM_148909.3
NP_683707.3
12
1099
NR_036662.1
11
990
XM_006710318.2
XP_006710381.1
11
829
XM_006710320.3
XP_006710383.1
9
666
XM_006710321.2
XP_006710384.1
8
662
XM_006710322.3
XP_006710385.1
9
823
XM_006710323.2
XP_006710386.1
8
731
XM_006710324.3
XP_006710387.1
10
741
XM_006710325.3
XP_006710388.1
9
711
XM_006710326.2
XP_006710389.1
7
713
XM_011540599.2
XP_011538901.1
13
923
XM_011540600.2
XP_011538902.1
12
890
XM_011540601.2
XP_011538903.1
11
851
XM_011540602.2
XP_011538904.1
12
825
XM_011540603.2
XP_011538905.1
12
849
XM_011540604.2
XP_011538906.1
9
624
XM_017000218.1
XP_016855707.1
10
790
XM_017000219.1
XP_016855708.1
11
810
XM_017000220.1
XP_016855709.1
8
551
XM_017000221.1
XP_016855710.1
8
502
XM_017000222.1
XP_016855711.1
12
1035
XM_017000223.1
XP_016855712.1
12
1033
XM_017000224.1
XP_016855713.1
9
978
XM_017000225.1
XP_016855714.1
10
829
GenBank mRNA
AB208851.1
-
1
336
AF392445.1
-
12
843
AK022554.1
-
7
470
AK027535.1
-
11
800
AK027707.1
-
8
626
AK056617.1
-
12
888
AK091703.1
-
11
772
AK124037.1
-
3
1316
AK128043.1
-
14
1293
AK297656.1
-
9
816
AK315167.1
-
8
662
AY178997.1
-
11
792
BC025978.2
-
8
696
3621193
CR457288.1
-
5
337
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0