Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYYR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320768.1 | NP_001307697.1 | 3 | 652 | ||
NM_052954.3 | NP_443186.2 | 3 | 652 | |||
NR_135472.1 | 4 | 819 | ||||
XM_011529450.2 | XP_011527752.1 | 3 | 532 | |||
GenBank mRNA | AF401639.1 | - | 3 | 257 | ||
AK223576.1 | - | 3 | 369 | |||
AL833200.1 | - | 3 | 463 | |||
AY061853.1 | - | 3 | 197 | |||
BC036761.1 | - | 3 | 440 | 5588436 | ||
JF693912.1 | - | 3 | 200 |