Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LYPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077427.3 | NP_001070895.1 | 2 | 480 | ||
NM_001321234.1 | NP_001308163.1 | 3 | 442 | |||
NM_001321235.1 | NP_001308164.1 | 2 | 603 | |||
NM_144586.6 | NP_653187.3 | 2 | 569 | |||
XM_011510546.2 | XP_011508848.1 | 2 | 1122 | |||
GenBank mRNA | AK075487.1 | - | 2 | 404 | ||
AK094501.1 | - | 3 | 345 | |||
AK123029.1 | - | 3 | 302 | |||
AK130005.1 | - | 3 | 281 | |||
AK309668.1 | - | 2 | 480 | |||
AK309849.1 | - | 3 | 289 | |||
AY358628.1 | - | 2 | 251 | |||
BC017318.2 | - | 2 | 382 | 3641660 | ||
BE386060.1 | - | 2 | 326 | |||
BP276862.1 | - | 2 | 433 | |||
CB110132.1 | - | 2 | 398 | |||
DA776269.1 | - | 2 | 471 |