Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM129A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052966.3 | NP_443198.1 | 5 | 728 | ||
XM_011509140.2 | XP_011507442.1 | 6 | 1603 | |||
XM_011509141.2 | XP_011507443.1 | 6 | 1603 | |||
GenBank mRNA | AB050477.1 | - | 5 | 654 | ||
AF288391.1 | - | 5 | 692 | |||
AK022527.1 | - | 5 | 566 | |||
AY550972.1 | - | 5 | 567 | |||
BC030531.1 | - | 5 | 584 | 5212338 |