Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC18B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052831.2 | NP_439896.1 | 9 | 1123 | ||
XM_017010236.1 | XP_016865725.1 | 9 | 1314 | |||
GenBank mRNA | AJ420537.1 | - | 3 | 249 | ||
AK124442.1 | - | 9 | 1002 | |||
AK289918.1 | - | 9 | 1142 | |||
AK293853.1 | - | 8 | 929 | |||
BC069567.1 | - | 5 | 526 | |||
BC069570.1 | - | 9 | 1043 | 7262298 |