Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054113.3 | NP_473454.1 | 3 | 211 | ||
GenBank mRNA | AB050868.1 | - | 3 | 161 | ||
BC069524.1 | - | 3 | 142 | |||
BC112200.1 | - | 3 | 211 | 8327668 | ||
BC113591.1 | - | 3 | 211 | 8322643 |