Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054113.3 | NP_473454.1 | 3, 4 | 239 | ||
GenBank mRNA | AB050868.1 | - | 3, 4 | 189 | ||
AW295492.1 | - | 3, 4 | 483 | |||
BC069524.1 | - | 3, 4 | 170 | |||
BC112200.1 | - | 3, 4 | 239 | 8327668 | ||
BC113591.1 | - | 3, 4 | 239 | 8322643 |