Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054114.4 | NP_473455.2 | 3 | 383 | ||
NM_138810.3 | NP_620165.1 | 3 | 383 | |||
NM_152133.2 | NP_687034.1 | 3 | 383 | |||
GenBank mRNA | AF385429.1 | - | 3 | 383 | ||
AF385430.1 | - | 3 | 383 | |||
AK097090.1 | - | 3 | 249 | |||
AL832662.1 | - | 3 | 259 | |||
BC015859.1 | - | 3 | 253 | |||
BC111731.1 | - | 3 | 239 | 40035199 |