Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114176.1 | NP_001107648.1 | 2 | 835 | ||
NM_001267043.1 | NP_001253972.1 | 2 | 285 | |||
NM_153207.4 | NP_694939.2 | 2 | 835 | |||
XM_006719031.3 | XP_006719094.1 | 2 | 1017 | |||
XM_011520560.2 | XP_011518862.1 | 2 | 1017 | |||
XM_017018806.1 | XP_016874295.1 | 2 | 162 | |||
GenBank mRNA | AB209384.1 | - | 2 | 629 | ||
AK126973.1 | - | 3 | 621 | |||
AK131410.1 | - | 2 | 285 | |||
BC015624.2 | - | 2 | 322 | 4843866 | ||
BC022220.1 | - | 2 | 810 | 3914323 |