Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139015.4 | NP_620584.2 | 8, 9 | 1264 | ||
XM_011537925.2 | XP_011536227.1 | 8, 9 | 794 | |||
GenBank mRNA | AB252457.1 | - | 8, 9 | 856 | ||
AJ345030.1 | - | 8, 9 | 832 | |||
AJ420898.1 | - | 8, 9 | 803 | |||
AK074916.1 | - | 7, 8 | 747 | |||
AK223590.1 | - | 8, 9 | 1260 | |||
AK291161.1 | - | 8, 9 | 1086 | |||
AY169313.1 | - | 8, 9 | 1038 | |||
AY358767.1 | - | 1, 2 | 110 | |||
BC009551.1 | - | 1, 2 | 132 | |||
BC025781.1 | - | 5, 6 | 539 | |||
BC073910.1 | - | 8, 9 | 1045 | 4538522 | ||
BC101625.1 | - | 8, 9 | 859 | 8069131 | ||
BC101627.1 | - | 8, 9 | 859 | 8069133 |