Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYKK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013619.3 | NP_001013641.2 | 3 | 545 | ||
NM_001083612.1 | NP_001077081.1 | 3 | 545 | |||
XM_011521231.2 | XP_011519533.1 | 3 | 527 | |||
XM_017021922.1 | XP_016877411.1 | 3 | 527 | |||
GenBank mRNA | AK126453.1 | - | 3 | 527 | ||
BC132753.1 | - | 3 | 492 | 40146775 | ||
BC144383.1 | - | 3 | 492 | 9052904 | ||
BM826406.1 | - | 2 | 234 | |||
DN831396.1 | - | 2 | 241 |