Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242929.1 | NP_001229858.1 | 2 | 265 | ||
NM_001308089.1 | NP_001295018.1 | 3 | 333 | |||
NM_001324130.1 | NP_001311059.1 | 2 | 268 | |||
NM_133450.3 | NP_597707.1 | 3 | 336 | |||
NR_040252.1 | 2 | 265 | ||||
XM_005255119.3 | XP_005255176.1 | 2 | 235 | |||
XM_005255121.3 | XP_005255178.1 | 2 | 137 | |||
XM_011522372.1 | XP_011520674.1 | 2 | 266 | |||
XM_011522373.1 | XP_011520675.1 | 2 | 148 | |||
XM_011522374.1 | XP_011520676.1 | 3 | 203 | |||
XM_011522378.1 | XP_011520680.1 | 2 | 92 | |||
XM_017022935.1 | XP_016878424.1 | 2 | 265 | |||
XM_017022936.1 | XP_016878425.1 | 2 | 268 | |||
GenBank mRNA | AK123371.1 | - | 2 | 136 | ||
AK123496.1 | - | 3 | 337 | |||
BC031350.1 | - | 2 | 88 | |||
BC050387.1 | - | 3 | 174 | 5751728 | ||
BM545219.1 | - | 2 | 111 | |||
BX333650.2 | - | 2 | 110 | |||
DR004376.1 | - | 2 | 271 |